2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals
2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 115821000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namehydrogen (2-chloroethyl)dimethylamine chloride
Molecular FormulaC4H11Cl2N
InChI KeyLQLJZSJKRYTKTP-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCCl
Molecular Weight (g/mol)144.04
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Chloride (Cl)24.1 to 25.1 % (ionic chloride)
Appearance (Color)White to brown-yellow
Melting point203°C to 210°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only