Thermo Scientific Chemicals

Dansyl Chloride 98%, Thermo Scientific Chemicals

Catalog number: 115850050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Dansyl Chloride 98%, Thermo Scientific Chemicals

Catalog number: 115850050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115850050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
605-65-2
IUPAC Name
5-(dimethylamino)naphthalene-1-sulfonyl chloride
Molecular Formula
C12H12ClNO2S
InChI Key
XPDXVDYUQZHFPV-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O
Appearance (Form)
Crystalline powder and/or chunks
Melting point
70°C to 74°C
Appearance (Color)
Yellow to light orange
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text