1-Dimethylamino-2-propanol, 99%, Thermo Scientific Chemicals
1-Dimethylamino-2-propanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Dimethylamino-2-propanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number 115870010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-16-7
IUPAC Name1-(dimethylamino)propan-2-ol
Molecular FormulaC5H13NO
InChI KeyNCXUNZWLEYGQAH-UHFFFAOYNA-N
SMILESCC(O)CN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
AppearanceMay discolor to yellow on storage
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only