3-Dimethylaminopropylamine, 99%, Thermo Scientific Chemicals
3-Dimethylaminopropylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Dimethylaminopropylamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
5 L
Packaging:
View all
Glass bottle
Catalog number 115900010
Price (USD)/ Each
42.65
Online exclusive
47.30 
Save 4.65 (10%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
42.65
Online exclusive
47.30 
Save 4.65 (10%)
Add to cart
3-Dimethylaminopropylamine, 99%, Thermo Scientific Chemicals
Catalog number115900010
Price (USD)/ Each
42.65
Online exclusive
47.30 
Save 4.65 (10%)
-
Add to cart
Chemical Identifiers
CAS109-55-7
IUPAC Name(3-aminopropyl)dimethylamine
Molecular FormulaC5H14N2
InChI KeyIUNMPGNGSSIWFP-UHFFFAOYSA-N
SMILESCN(C)CCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4340 to 1.4360 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<15 APHA
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only