Thermo Scientific Chemicals

3-Dimethylaminopropylchloride hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 115915000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Dimethylaminopropylchloride hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 115915000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115915000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5407-04-5
IUPAC Name
hydrogen (3-chloropropyl)dimethylamine chloride
Molecular Formula
C5H13Cl2N
InChI Key
LJQNMDZRCXJETK-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)CCCCl
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Loss on drying
=<1 % (105°C, 3 hrs)
Chloride (Cl)
21.9 to 22.9 % (titr.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text