N,N-Dimethylaniline, 99%, Thermo Scientific Chemicals
N,N-Dimethylaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylaniline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 115920010
Price (USD)/ Each
47.65
Online exclusive
52.40 
Save 4.75 (9%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
52.40 
Save 4.75 (9%)
Add to cart
N,N-Dimethylaniline, 99%, Thermo Scientific Chemicals
Catalog number115920010
Price (USD)/ Each
47.65
Online exclusive
52.40 
Save 4.75 (9%)
-
Add to cart
Chemical Identifiers
CAS121-69-7
IUPAC NameN,N-dimethylaniline
Molecular FormulaC8H11N
InChI KeyJLTDJTHDQAWBAV-UHFFFAOYSA-N
SMILESCN(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5570 to 1.5590 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only