Thermo Scientific Chemicals

2,6-Dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: 115945000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

2,6-Dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: 115945000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115945000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
87-62-7
IUPAC Name
2,6-dimethylaniline
Molecular Formula
C8H11N
InChI Key
UFFBMTHBGFGIHF-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C)=C1N
GC
>=98.5 %
Refractive index
1.5591 to 1.5611 (20°C, 589 nm)
Appearance (Color)
Clear yellow to red-brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text