3,4-Dimethylbenzoic acid, 98%, Thermo Scientific Chemicals
3,4-Dimethylbenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 115990100
Price (USD)/ Each
26.80
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
26.80
Add to cart
3,4-Dimethylbenzoic acid, 98%, Thermo Scientific Chemicals
Catalog number115990100
Price (USD)/ Each
26.80
-
Add to cart
Chemical Identifiers
CAS619-04-5
IUPAC Name3,4-dimethylbenzoate
Molecular FormulaC9H9O2
InChI KeyOPVAJFQBSDUNQA-UHFFFAOYSA-M
SMILESCC1=CC=C(C=C1C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point162°C to 168°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only