Thermo Scientific Chemicals

5,6-Dimethyl-1H-benzotriazole hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 116050250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

5,6-Dimethyl-1H-benzotriazole hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 116050250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 116050250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4184-79-6
IUPAC Name
5,6-dimethyl-1H-1,2,3-benzotriazole
Molecular Formula
C8H9N3
InChI Key
MVPKIPGHRNIOPT-UHFFFAOYSA-N
SMILES
CC1=C(C)C=C2N=NNC2=C1
Water
=<11 % (K.F.)
Titration with TBAH
>=98.5 % (on dry substance)
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
150°C to 156°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text