5,5-Dimethyl-1,3-cyclohexanedione, 99%, Thermo Scientific Chemicals
5,5-Dimethyl-1,3-cyclohexanedione, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,5-Dimethyl-1,3-cyclohexanedione, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 116150250
Price (USD)/ Each
32.65
Online exclusive
36.00 
Save 3.35 (9%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
32.65
Online exclusive
36.00 
Save 3.35 (9%)
Add to cart
5,5-Dimethyl-1,3-cyclohexanedione, 99%, Thermo Scientific Chemicals
Catalog number116150250
Price (USD)/ Each
32.65
Online exclusive
36.00 
Save 3.35 (9%)
-
Add to cart
Chemical Identifiers
CAS126-81-8
IUPAC Name5,5-dimethylcyclohexane-1,3-dione
Molecular FormulaC8H12O2
InChI KeyBADXJIPKFRBFOT-UHFFFAOYSA-N
SMILESCC1(C)CC(=O)CC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point146°C to 150°C
HPLC>=98.5 %
Appearance (Color)White to yellow to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only