Thermo Scientific Chemicals

5,5-Dimethyl-1,3-cyclohexanedione, 99%, Thermo Scientific Chemicals

Catalog number: 116151000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

5,5-Dimethyl-1,3-cyclohexanedione, 99%, Thermo Scientific Chemicals

Catalog number: 116151000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 116151000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
126-81-8
IUPAC Name
5,5-dimethylcyclohexane-1,3-dione
Molecular Formula
C8H12O2
InChI Key
BADXJIPKFRBFOT-UHFFFAOYSA-N
SMILES
CC1(C)CC(=O)CC(=O)C1
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
146°C to 150°C
HPLC
>=98.5 %
Appearance (Color)
White to yellow to light beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text