unsym-Dimethylethylenediamine, 99%, Thermo Scientific Chemicals
unsym-Dimethylethylenediamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

unsym-Dimethylethylenediamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 116200250
Price (USD)/ Each
50.20
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
50.20
Add to cart
unsym-Dimethylethylenediamine, 99%, Thermo Scientific Chemicals
Catalog number116200250
Price (USD)/ Each
50.20
-
Add to cart
Chemical Identifiers
CAS108-00-9
IUPAC Name(2-aminoethyl)dimethylamine
Molecular FormulaC4H12N2
InChI KeyDILRJUIACXKSQE-UHFFFAOYSA-N
SMILESCN(C)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Water=<1 %
Refractive index1.4255 to 1.4275 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only