2,4-Dimethyl-3-ethylpyrrole, 96%, Thermo Scientific Chemicals
2,4-Dimethyl-3-ethylpyrrole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dimethyl-3-ethylpyrrole, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 116210050
Price (USD)/ Each
110.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
130.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
110.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
130.00 
Save 19.35 (15%)
Add to cart
2,4-Dimethyl-3-ethylpyrrole, 96%, Thermo Scientific Chemicals
Catalog number116210050
Price (USD)/ Each
110.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
130.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
CAS517-22-6
IUPAC Name3-ethyl-2,4-dimethyl-1H-pyrrole
Molecular FormulaC8H13N
InChI KeyZEBBLOXDLGIMEG-UHFFFAOYSA-N
SMILESCCC1=C(C)NC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to red-brown
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=95 %
Refractive index1.4960 to 1.4980 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only