Thermo Scientific Chemicals

Dimethyl glutarate, 98%, Thermo Scientific Chemicals

Catalog number: 116231000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethyl glutarate, 98%, Thermo Scientific Chemicals

Catalog number: 116231000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 116231000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1119-40-0
IUPAC Name
1,5-dimethyl pentanedioate
Molecular Formula
C7H12O4
InChI Key
XTDYIOOONNVFMA-UHFFFAOYSA-N
SMILES
COC(=O)CCCC(=O)OC
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4236 to 1.4256 (20°C, 589 nm)
Appearance (Color)
Clear colorless
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text