N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 116340050
Price (USD)/ Each
50.60
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
50.60
Add to cart
N,O-Dimethylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog number116340050
Price (USD)/ Each
50.60
-
Add to cart
Chemical Identifiers
CAS6638-79-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals
Titration with HClO4>=97.5 % (on anhydrous substance)
Water=<0.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only