2,3-Dimethylmaleic anhydride, 97%, Thermo Scientific Chemicals
2,3-Dimethylmaleic anhydride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dimethylmaleic anhydride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Packaging:
Glass bottle
Catalog number 116390050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS766-39-2
IUPAC Namedimethyl-2,5-dihydrofuran-2,5-dione
Molecular FormulaC6H6O3
InChI KeyMFGALGYVFGDXIX-UHFFFAOYSA-N
SMILESCC1=C(C)C(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Titration with TBAH>=96.0 %
Appearance (Color)White to beige
Melting point91°C to 96°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only