Thermo Scientific Chemicals

N,N-Dimethyl-3-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 116470250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Dimethyl-3-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 116470250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 116470250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
619-31-8
IUPAC Name
N,N-dimethyl-3-nitroaniline
Molecular Formula
C8H10N2O2
InChI Key
CJDICMLSLYHRPT-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC(=C1)[N+]([O-])=O
Appearance (Color)
Orange to orange-red
Titration with HClO4
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Melting point
57°C to 61°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text