4,5-Dimethyl-o-phenylenediamine, 98%, Thermo Scientific Chemicals
4,5-Dimethyl-o-phenylenediamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,5-Dimethyl-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 116610250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3171-45-7
IUPAC Name4,5-dimethylbenzene-1,2-diamine
Molecular FormulaC8H12N2
InChI KeyXSZYBMMYQCYIPC-UHFFFAOYSA-N
SMILESCC1=CC(N)=C(N)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
GC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder or solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only