Thermo Scientific Chemicals

Dimethyl Phthalate, 99%, Thermo Scientific Chemicals

Catalog number: 116640025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Dimethyl Phthalate, 99%, Thermo Scientific Chemicals

Catalog number: 116640025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 116640025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
131-11-3
IUPAC Name
1,2-dimethyl benzene-1,2-dicarboxylate
Molecular Formula
C10H10O4
InChI Key
NIQCNGHVCWTJSM-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1C(=O)OC
Color scale
=<20 APHA
GC
>=98.5 %
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms
Refractive index
1.5140 to 1.5160 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text