2,5-Dimethylpyrrole, 97%, Thermo Scientific Chemicals
2,5-Dimethylpyrrole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dimethylpyrrole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 116750250
Price (USD)/ Each
135.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
159.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
135.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
159.00 
Save 23.35 (15%)
Add to cart
2,5-Dimethylpyrrole, 97%, Thermo Scientific Chemicals
Catalog number116750250
Price (USD)/ Each
135.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
159.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,5-dimethyl-1H-pyrrole
Molecular FormulaC6H9N
InChI KeyPAPNRQCYSFBWDI-UHFFFAOYSA-N
SMILESCC1=CC=C(C)N1
Molecular Weight (g/mol)95.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
AppearanceMay darken on storage
Refractive index1.5040 to 1.5080 (20°C, 589 nm)
Appearance (Color)Clear yellow to orange to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only