Thermo Scientific Chemicals

N,N'-Dimethylthiourea, 99%, Thermo Scientific Chemicals

Catalog number: 116891000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N'-Dimethylthiourea, 99%, Thermo Scientific Chemicals

Catalog number: 116891000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 116891000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
534-13-4
IUPAC Name
1,3-dimethylthiourea
Molecular Formula
C3H8N2S
InChI Key
VLCDUOXHFNUCKK-UHFFFAOYSA-N
SMILES
CNC(=S)NC
Appearance (Form)
Crystalline powder or crystals and/or chunks
Melting point
61°C to 67°C
Appearance (Color)
Colorless to white
Titration with HClO4
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text