1,3-Dimethylurea, 98%, Thermo Scientific Chemicals
1,3-Dimethylurea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Dimethylurea, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
Plastic bottle
Catalog number 116910010
Price (USD)/ Each
66.70
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
66.70
Add to cart
1,3-Dimethylurea, 98%, Thermo Scientific Chemicals
Catalog number116910010
Price (USD)/ Each
66.70
-
Add to cart
Chemical Identifiers
CAS96-31-1
IUPAC Name1,3-dimethylurea
Molecular FormulaC3H8N2O
InChI KeyMGJKQDOBUOMPEZ-UHFFFAOYSA-N
SMILESCNC(=O)NC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)White
Melting point103°C to 108°C
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only