2,4-Dinitrofluorobenzene, 98%, Thermo Scientific Chemicals
2,4-Dinitrofluorobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dinitrofluorobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 117035000
Price (USD)/ Each
793.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
933.00 
Save 139.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
793.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
933.00 
Save 139.35 (15%)
Add to cart
2,4-Dinitrofluorobenzene, 98%, Thermo Scientific Chemicals
Catalog number117035000
Price (USD)/ Each
793.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
933.00 
Save 139.35 (15%)
-
Add to cart
Chemical Identifiers
CAS70-34-8
IUPAC Name1-fluoro-2,4-dinitrobenzene
Molecular FormulaC6H3FN2O4
InChI KeyLOTKRQAVGJMPNV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or low melting solid
GC>=97.5 %
Appearance (Color)Yellow to brown
Infrared spectrumConforms
Refractive index1.5675 to 1.5705 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only