Dioctyl sulfosuccinate, sodium salt, 96%, Thermo Scientific Chemicals
Dioctyl sulfosuccinate, sodium salt, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dioctyl sulfosuccinate, sodium salt, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 117101000
Price (USD)/ Each
44.65
Online exclusive
49.40 
Save 4.75 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
44.65
Online exclusive
49.40 
Save 4.75 (10%)
Add to cart
Dioctyl sulfosuccinate, sodium salt, 96%, Thermo Scientific Chemicals
Catalog number117101000
Price (USD)/ Each
44.65
Online exclusive
49.40 
Save 4.75 (10%)
-
Add to cart
Chemical Identifiers
CAS577-11-7
IUPAC Namesodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
Molecular FormulaC20H37NaO7S
InChI KeyAPSBXTVYXVQYAB-UHFFFAOYNA-M
SMILES[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Waxy solid
Assay>=95 %
Infrared spectrumConforms
Water=<2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only