1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 117110025
Price (USD)/ Each
194.65
Online exclusive
216.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
194.65
Online exclusive
216.00 
Save 21.35 (10%)
Add to cart
1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals
Catalog number117110025
Price (USD)/ Each
194.65
Online exclusive
216.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Peroxides=<0.005 % (as H2O2)
Appearance (Form)Clear liquid
Water=<0.1 %
Stabilizer5 to 30 ppm BHT
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only