Thermo Scientific Chemicals

Dipentaerythritol, 85+%, technical, Thermo Scientific Chemicals

Catalog number: 117120250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Dipentaerythritol, 85+%, technical, Thermo Scientific Chemicals

Catalog number: 117120250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 117120250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
126-58-9
IUPAC Name
2-{[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl}-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C10H22O7
InChI Key
TXBCBTDQIULDIA-UHFFFAOYSA-N
SMILES
OCC(CO)(CO)COCC(CO)(CO)CO
Appearance (Color)
White
Melting point
215°C to 225°C
Hydroxyl value
1125 to 1325 mg KOH/g
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text