Dipentaerythritol, 85+%, technical, Thermo Scientific Chemicals
Dipentaerythritol, 85+%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dipentaerythritol, 85+%, technical, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Packaging:
Plastic bottle
Catalog number 117125000
Price (USD)/ Each
67.10
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
67.10
Add to cart
Dipentaerythritol, 85+%, technical, Thermo Scientific Chemicals
Catalog number117125000
Price (USD)/ Each
67.10
-
Add to cart
Chemical Identifiers
CAS126-58-9
IUPAC Name2-{[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl}-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC10H22O7
InChI KeyTXBCBTDQIULDIA-UHFFFAOYSA-N
SMILESOCC(CO)(CO)COCC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point215°C to 225°C
Hydroxyl value1125 to 1325 mg KOH/g
Appearance (Form)Powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only