Diphenylacetic Acid, 99+%, Thermo Scientific Chemicals
Diphenylacetic Acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylacetic Acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 117150050
Price (USD)/ Each
18.65
Online exclusive
20.50 
Save 1.85 (9%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
18.65
Online exclusive
20.50 
Save 1.85 (9%)
Add to cart
Diphenylacetic Acid, 99+%, Thermo Scientific Chemicals
Catalog number117150050
Price (USD)/ Each
18.65
Online exclusive
20.50 
Save 1.85 (9%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,2-diphenylacetic acid
Molecular FormulaC14H12O2
InChI KeyPYHXGXCGESYPCW-UHFFFAOYSA-N
SMILESOC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)212.25
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Melting point146°C to 149°C
Appearance (Color)White to off-white to cream
Titration with NaOH>=99.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only