Thermo Scientific Chemicals

Diphenylacetic Acid, 99+%, Thermo Scientific Chemicals

Catalog number: 117151000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Diphenylacetic Acid, 99+%, Thermo Scientific Chemicals

Catalog number: 117151000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 117151000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
117-34-0
IUPAC Name
2,2-diphenylacetic acid
Molecular Formula
C14H12O2
InChI Key
PYHXGXCGESYPCW-UHFFFAOYSA-N
SMILES
OC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to off-white to cream
Titration with NaOH
>=99.0 %
Appearance (Form)
Powder or crystals and/or chunks
Melting point
146°C to 149°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text