1,3-Diphenylacetone, 99%, Thermo Scientific Chemicals
1,3-Diphenylacetone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Diphenylacetone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 117175000
Price (USD)/ Each
632.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
744.00 
Save 111.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
632.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
744.00 
Save 111.35 (15%)
Add to cart
1,3-Diphenylacetone, 99%, Thermo Scientific Chemicals
Catalog number117175000
Price (USD)/ Each
632.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
744.00 
Save 111.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,3-diphenylpropan-2-one
Molecular FormulaC15H14O
InChI KeyYFKBXYGUSOXJGS-UHFFFAOYSA-N
SMILESO=C(CC1=CC=CC=C1)CC1=CC=CC=C1
Molecular Weight (g/mol)210.28
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
Appearance (Form)Liquid or crystalline mass
Melting point30°C to 34°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only