Diphenylacetylene, 99%, Thermo Scientific Chemicals
Diphenylacetylene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylacetylene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 117181000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2-phenylethynyl)benzene
Molecular FormulaC14H10
InChI KeyJRXXLCKWQFKACW-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C#CC1=CC=CC=C1
Molecular Weight (g/mol)178.23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to brown
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder and/or chunks
Melting point58°C to 61°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only