2,2-Diphenylethylamine, 96%, Thermo Scientific Chemicals
2,2-Diphenylethylamine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Diphenylethylamine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 117250250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3963-62-0
IUPAC Name2,2-diphenylethan-1-amine
Molecular FormulaC14H15N
InChI KeyRXMTUVIKZRXSSM-UHFFFAOYSA-N
SMILESNCC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Solid
Melting point43°C to 49°C
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only