Invitrogen™

SuperScript™ III Platinum™ One-Step qRT-PCR Kit

Catalog number: 11732088
Have Questions?
Invitrogen™

SuperScript™ III Platinum™ One-Step qRT-PCR Kit

Catalog number: 11732088
Have Questions?
Catalog Number
11732088
Unit Size
500 reactions
Price (USD)
Quantity
Catalog Number
Unit Size
100 reactions
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
11732088500 reactions
11732020100 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The SuperScript III Platinum One-Step qRT-PCR Kit’s proven formula delivers reliable, accurate target detection when it matters most. The convenient one-step format and consistent performance make it ideal for a number of gene expression applications, including infectious disease surveillance. The kit has been validated by several global public health organizations and has long been the choice of the US CDC for infectious disease surveillance.

Features of the SuperScript III Platinum One-Step qRT-PCR Kit include:
• SuperScript III RT for higher temperature cDNA synthesis when addressing difficult RNA secondary structure
• Platinum Taq DNA Polymerase with hot-start technology for improved specificity
• LUX fluorogenic primers and dual-labeled fluorogenic probes for effective detection performance
• Validated performance on multiple real-time instrument platforms, including rotor-based systems

SuperScript III Platinum One-Step qRT-PCR Kits come in multiple configurations:
SuperScript III Platinum One-Step qRT-PCR Kit (this page)
SuperScript III Platinum One-Step qRT-PCR Kit with ROX passive reference
SuperScript III Platinum SYBR Green One-Step qRT-PCR Kit
SuperScript III Platinum SYBR Green One-Step qRT-PCR Kit with ROX passive reference

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
PCR Method
1-step RT-qPCR
Polymerase
Taq DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ
Product Line
Platinum™, SuperScript™
Product Type
One-Step qRT-PCR Kit
Shipping Condition
Dry Ice
Sufficient For
500 Reactions
For Use With (Application)
Gene Expression, Pathogen Detection, Virus Detection
No. of Reactions
500 Reactions
Sample Type
RNA

Contents & Storage

• 500 μl SuperScript™ III/Platinum™ Taq Mix
• 12.5 ml 2X Reaction Mix
• 2 × 1 ml Magnesium Sulfate (50 mM)
• 500 μl ROX Reference Dye (25 μM)

Store components at -20°C. ROX Reference Dye must be stored in the dark.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text