Invitrogen™

Platinum™ SYBR™ Green qPCR SuperMix-UDG

Catalog number: 11733038
Have Questions?
Invitrogen™

Platinum™ SYBR™ Green qPCR SuperMix-UDG

Catalog number: 11733038
Have Questions?
Catalog Number
11733038
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
11733038100 reactions
-
11733046500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of Platinum SYBR Green qPCR SuperMix-UDG and added additional capabilities for your gene expression analysis.

**********************
Platinum™ SYBR™ Green qPCR SuperMix-UDG provides a sensitive and convenient system for real-time PCR detection with SYBR™ Green I dye. The SuperMix format combines amplification and detection reagents into a ready-to-go mix to make set-up easy and ensure reproducible results. Platinum™ SYBR™ Green qPCR SuperMix-UDG offers:

–Platinum™ Taq DNA Polymerase for sensitive amplification from as few as 10 copies of target (Figure 1)

–100% dUTP and UDG enzyme for superior control of carryover contamination, saving time by reducing the number of failed experiments
–Separate tubes of ROX reference dye and BSA for easy optimization on 96-well plates and glass capillary tube instruments (Figure 2)
–SYBR™ Green I dye for simple and easy detection of real-time product
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
SYBR
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Taq DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, GeneAmp 5700, StepOne™, StepOnePlus™, ViiA™ 7 System, Cepheid SmartCycler, BioRad iCycler iQ
Product Line
Platinum™, SYBR™
Product Type
Real Time PCR SYBR Master Mix
Shipping Condition
Dry Ice
Sufficient For
100 Reactions of 50 μL Each
For Use With (Application)
Gene Expression
No. of Reactions
100 Reactions
Sample Type
DNA (Genomic), cDNA
Volume
2 x 1.25 mL

Contents & Storage

• Platinum™ SYBR™ Green qPCR SuperMix-UDG (2 × 1.25 mL)
• 50 mM Magnesium Chloride (1 mL)
• ROX Reference Dye (100 μL)
• 20X BSA (300 μL)

Sufficient reagents are provided for 100 reactions of 50 μL each. Store all components at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text