Diphenylmethane, 99%, Thermo Scientific Chemicals
Diphenylmethane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylmethane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Catalog number 117355000
Price (USD)/ Each
68.10
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
68.10
Add to cart
Diphenylmethane, 99%, Thermo Scientific Chemicals
Catalog number117355000
Price (USD)/ Each
68.10
-
Add to cart
Chemical Identifiers
CAS101-81-5
IUPAC Namebenzylbenzene
Molecular FormulaC13H12
InChI KeyCZZYITDELCSZES-UHFFFAOYSA-N
SMILESC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
GC>=98.5 %
Appearance (Color)Colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only