Thermo Scientific Chemicals

2,5-Diphenyloxazole, Scintillation Grade 99%, Thermo Scientific Chemicals

Catalog number: 117380250
25 g, Plastic Bottle, Each
Thermo Scientific Chemicals

2,5-Diphenyloxazole, Scintillation Grade 99%, Thermo Scientific Chemicals

Catalog number: 117380250
25 g, Plastic Bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 117380250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92-71-7
IUPAC Name
2,5-diphenyl-1,3-oxazole
Molecular Formula
C15H11NO
InChI Key
CNRNYORZJGVOSY-UHFFFAOYSA-N
SMILES
O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1
Melting point
70°C to 72°C
UV transmittance
>=85 % (40% in toluene)
Appearance (Color)
White
Infrared spectrum
Conforms
Color of solution
=<50 APHA (40% in toluene)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for fluorescence/luminescence spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text