Thermo Scientific Chemicals

Diphenyl Phosphite, Contains Varying Amounts of Phenol and (C6H5O)3P, Thermo Scientific Chemicals

Catalog number: 117390050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Diphenyl Phosphite, Contains Varying Amounts of Phenol and (C6H5O)3P, Thermo Scientific Chemicals

Catalog number: 117390050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 117390050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4712-55-4
IUPAC Name
oxodiphenoxy-λ⁵-phosphanylium
Molecular Formula
C12H10O3P
InChI Key
CDXVUROVRIFQMV-UHFFFAOYSA-N
SMILES
O=[P+](OC1=CC=CC=C1)OC1=CC=CC=C1
Infrared spectrum
Conforms
Acid value
=<15 mg KOH/g
Color scale
=<50 APHA
Refractive index
1.5565 to 1.5585 (20°C, 589 nm)
Specific gravity
(25/15.5°C) 1.2 to 1.22

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text