2,2'-Dipyridyl, 99+%, Thermo Scientific Chemicals
2,2'-Dipyridyl, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2'-Dipyridyl, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 117505000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS366-18-7
IUPAC Name2,2'-bipyridine
Molecular FormulaC10H8N2
InChI KeyROFVEXUMMXZLPA-UHFFFAOYSA-N
SMILESC1=CC=C(N=C1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point69°C to 72°C
Suitability(for Fe determination) Passes test
Appearance (Form)Crystalline powder or crystals
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only