SuperScript™ IV Single Cell/Low Input cDNA PreAmp Kit
SuperScript™ IV Single Cell/Low Input cDNA PreAmp Kit
Invitrogen™

SuperScript™ IV Single Cell/Low Input cDNA PreAmp Kit

The Invitrogen SuperScript IV Single Cell/Low Input cDNA PreAmp Kit is designed for efficient cDNA synthesis and amplification directly fromRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
1175204848 Reactions
1175209696 Reactions
11752192192 Reactions
11752384384 Reactions
11752480480 Reactions
Catalog number 11752048
Price (KRW)
1,758,000
Online offer
Ends: 30-Jun-2025
2,000,000
Save 242,000 (12%)
Each
-
Add to cart
No. of Reactions:
48 Reactions
Recurring order eligible. Learn more »
Request bulk or custom format
Price (KRW)
1,758,000
Online offer
Ends: 30-Jun-2025
2,000,000
Save 242,000 (12%)
Each
Add to cart
Ask our AI about this Product

The Invitrogen SuperScript IV Single Cell/Low Input cDNA PreAmp Kit is designed for efficient cDNA synthesis and amplification directly from intact single cells (1–1,000) or low amounts of total RNA (2 pg–10 ng). It contains all required components to perform cell lysis, reverse transcription (RT), and PCR amplification in a convenient premixed format. A combination of superior enzymes (SuperScript IV Reverse Transcriptase and Platinum SuperFi U DNA Polymerase) enables high yields, sensitivity, and accuracy. Due to the short reaction times and minimal hands-on time, this simple one-tube preamplification protocol can be completed in just over two hours. The high quality global cDNA obtained can be used for next-generation sequencing (NGS) applications or for gene expression analysis by real-time PCR.

Features of the SuperScript IV Single Cell/Low Input cDNA PreAmp Kit include:
• Superior sensitivity for easy detection of low abundant targets (as low as single cell or 2 pg total RNA)
• High quality cDNA for full-length transcript information with uniform coverage
• Streamlined workflow with simple one-tube protocol
• Short lysis and reverse transcription times for time savings
• Global preamplification compatible with downstream analysis by NGS or real-time PCR

The SuperScript IV Single Cell/Low Input cDNA PreAmp Kit leverages the terminal deoxynucleotidyl transferase (TdT) activity of the SuperScript IV Reverse Transcriptase. Capturing oligonucleotide containing oligo(dT) and adapter sequences is used as primer for selective first strand cDNA synthesis from poly(A)-containing RNAs. When the reverse transcriptase reaches the 5‘ end of the RNA template, TdT activity adds 1–3 extra nucleotides to the cDNA end, enabling template switching and ligation-free incorporation of adapter sequence in the 3‘ end of the resulting cDNA. The high sensitivity and template switching efficiency of SuperScript IV Reverse Transcriptase allow the capture of even low-abundance targets.

Adapter sequences incorporated in both ends of the cDNA serve as primer-binding sites in the subsequent PCR amplification step, allowing global preamplification of the full-length templates. The included Platinum SuperFi U Preamplification Master Mix contains a superior high-fidelity DNA polymerase that ensures efficient amplification of long templates (up to 10 kb) with no PCR-induced bias or errors.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypePCR Amplified cDNA
FormatKit
No. of Reactions48 Reactions
Optimal Reaction Temperature50°C
PolymerasePlatinum SuperFi U DNA Polymerase
Quantity48 reactions
Reagent TypeReverse Transcription
Reverse TranscriptaseSuperScript IV
Shipping ConditionDry Ice
Size (Final Product)Up to 10 kb
Starting MaterialCells or RNA
TechniqueReverse Transcription, Preamplification
For Use With (Application)Next-Generation Sequencing (NGS), Real Time PCR (qPCR)
GC-Rich PCR PerformanceHigh
Reaction Speed30 min.
Unit SizeEach
Contents & Storage

• 10X Lysis Buffer (48 μL)
• RNase Inhibitor (19 μL)
• Capturing Oligonucleotide (48 μL)
• 4X SuperScript IV cDNA Synthesis Master Mix (240 μL)
• Template Switching Oligonucleotide (48 μL)
• 2X Platinum SuperFi U Preamplification Master Mix (2 x 1.2 mL)
• Preamplification primer (48 μL)
• Nuclease-free water (2 x 1.25 mL)

Store at –15 to –25°C.

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3265906Certificate of AnalysisJun 20, 202511752048
3255661Certificate of AnalysisJun 06, 202511752048
3255644Certificate of AnalysisJun 06, 202511752048
3255544Certificate of AnalysisJun 06, 202511752048
3178976Certificate of AnalysisFeb 26, 202511752048
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

We recommend using TaqMan Fast Advanced Master Mix for all TaqMan‑based detection methods.

We recommend using PowerTrack SYBR Green Master Mix for all SYBR Green‑based detection methods.

Cycle number depends on sample type and input amount.
For example:
- 21 cycles for 2 pg of total RNA
- 18 cycles for 10 pg of total RNA, or single cells
- 15 cycles for 100 pg of total RNA, or up to 10 cells
- 11 cycles for 1-10 ng of total RNA, or 100–1,000 cells
- RNA quantity in cells can vary by cell type, cell cycle, and cell health. Optimization of the PCR cycle number may be needed to reach desired yields. An additional 2–3 cycles are required for smaller cells, such as Jurkat, Daudi, or peripheral blood mononuclear cells (PBMC).

The master mix includes Platinum SuperFi U DNA Polymerase. It provides the same benefits as Platinum SuperFi II DNA Polymerase, but is also compatible with uracil.

Lysis buffer is included in the kit, therefore intact mammalian cells can be used. Purification of total RNA is required for cDNA preamplification of RNA isolated from plant, fungi, or other cell wall‑containing organisms.

rRNA depletion or mRNA enrichment is not required, because only poly(A)‑containing RNA is amplified.

Share catalog number, name or link

1x1 image pixel for data collection