Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

CellsDirect™ One-Step qRT-PCR Kit with ROX, for ABI 7000, 7300, 7700, and 7900(HT) instruments

Catalog number: 11754100
Have Questions?
Invitrogen™

CellsDirect™ One-Step qRT-PCR Kit with ROX, for ABI 7000, 7300, 7700, and 7900(HT) instruments

Catalog number: 11754100
Have Questions?
Catalog Number
11754100
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
11754100100 reactions
-
11754500500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The CellsDirect™ One-Step qRT-PCR Kit delivers highly sensitive and specific real-time qPCR results directly from cells, without the need for an RNA purification step.

Features of the CellsDirect™ One-Step qRT-PCR Kit include:
Efficiency—Offers significant time and cost savings for increased throughput
Convenience—Enzyme mix includes both SuperScript™ III Reverse Transcriptase and Platinum™ Taq DNA Polymerase
Sensitivity—No separate RNA isolation step increases sensitivity by minimizing sample loss
Broad Detection Range—Detect targets from samples ranging from a single cell to 10,000 cells
Versatility—Compatible with a wide variety of cell lines and samples collected by Laser Capture Microdissection (LCM)

Streamline Your Workflow
In traditional real-time quantitative RT-PCR (qRT-PCR), you first isolate RNA from cells in a time-consuming procedure that can lead to loss of material. Using the CellsDirect™ One-Step qRT-PCR Kit, you lyse the cells and add the complete lysate directly to a one-step qRT-PCR reaction with minimal handling and sample loss.

Ideal for Your qRT-PCR Application
CellsDirect™ technology is validated for gene expression studies involving limited samples, and is suitable for validation of RNAi knockdowns. CellsDirect™ One-Step qRT-PCR Kits are especially valuable for high-throughput applications by eliminating costly, time-consuming RNA purification steps (Figure 1). The CellsDirect™ One-Step qRT-PCR Kit may also be used to detect genomic DNA targets.

CellsDirect™ One-Step qRT-PCR Kits may be used in conjunction with hydrolysis probes (e.g. TaqMan™) or LUX™ fluorogenic primers.

Note: This kit includes 2X Reaction Mix containing ROX™ Reference Dye. An equivalent kit (11753-500), containing a separate tube of ROX™ Reference Dye, is also available.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Reaction Speed
Fast or Standard
For Use With (Equipment)
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, QuantStudio 3 and 5 systems, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Product Line
CellsDirect™, Platinum™, SuperScript™
For Use With (Application)
RT-PCR
Concentration
2X
No. of Reactions
100 Reactions
Sample Type
Cells

Contents & Storage

• Resuspension Buffer, 10 mL
• Lysis Enhancer, 1 mL
• 1 U/μL DNase I, Amplification Grade, 500 μL
• 10X DNase I Buffer, 160 μL
• 25 mM EDTA, 400 μL
• SuperScript™ III RT/Platinum™ Taq Mix, 100 μL
• 2X Reaction Mix with ROX™, 2 x 1.25 mL
• 50 mM MgSO4, 1 mL
• DEPC-treated water, 2 mL
• 10 ng/μL HeLa Total RNA, 10 μL

The kit is shipped on dry ice and should be stored at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text