Thermo Scientific Chemicals

Dithiooxamide, 98%, Thermo Scientific Chemicals

Catalog number: 117551000
100 g, Each
Thermo Scientific Chemicals

Dithiooxamide, 98%, Thermo Scientific Chemicals

Catalog number: 117551000
100 g, Each
Quantity
Catalog number: 117551000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-40-3
IUPAC Name
ethanedithioamide
Molecular Formula
C2H4N2S2
InChI Key
OAEGRYMCJYIXQT-UHFFFAOYSA-N
SMILES
NC(=S)C(N)=S
Appearance (Color)
Orange to red to brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Elemental Analysis
19.5 to 20.5 % (C)
Sulfated ash
=<2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text