Thermo Scientific Chemicals

(-)-Di-p-toluoyl-L-tartaric acid, 97%, Thermo Scientific Chemicals

Catalog number: 117570250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

(-)-Di-p-toluoyl-L-tartaric acid, 97%, Thermo Scientific Chemicals

Catalog number: 117570250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 117570250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
32634-66-5
IUPAC Name
(2R,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Molecular Formula
C20H18O8
InChI Key
CMIBUZBMZCBCAT-IYBDPMFKSA-N
SMILES
CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
165°C to 171°C
Specific optical rotation
-139° to -145° (20°C, 589 nm) (c=1, ethanol)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text