Thermo Scientific Chemicals

n-Dodecane, 99%, pure, Thermo Scientific Chemicals

Catalog number: 117590100
10 L, Metal Drum, Each
Thermo Scientific Chemicals

n-Dodecane, 99%, pure, Thermo Scientific Chemicals

Catalog number: 117590100
10 L, Metal Drum, Each
Quantity
Packaging
Catalog number: 117590100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
112-40-3
IUPAC Name
dodecane
Molecular Formula
C12H26
InChI Key
SNRUBQQJIBEYMU-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4210 to 1.4230 (20°C, 589 nm)
Color scale
=<10 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text