n-Dodecane, 99%, pure, Thermo Scientific Chemicals
n-Dodecane, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Dodecane, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
100 mL
500 mL
2.5 L
10 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 117592500
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
Add to cart
n-Dodecane, 99%, pure, Thermo Scientific Chemicals
Catalog number117592500
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
133.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
CAS112-40-3
IUPAC Namedodecane
Molecular FormulaC12H26
InChI KeySNRUBQQJIBEYMU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4210 to 1.4230 (20°C, 589 nm)
Color scale=<10 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only