1,12-Dodecanediol, 98%, Thermo Scientific Chemicals
1,12-Dodecanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,12-Dodecanediol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number 117611000
Price (USD)/ Each
53.65
Online exclusive
63.40 
Save 9.75 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
53.65
Online exclusive
63.40 
Save 9.75 (15%)
Add to cart
1,12-Dodecanediol, 98%, Thermo Scientific Chemicals
Catalog number117611000
Price (USD)/ Each
53.65
Online exclusive
63.40 
Save 9.75 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedodecane-1,12-diol
Molecular FormulaC12H26O2
InChI KeyGHLKSLMMWAKNBM-UHFFFAOYSA-N
SMILESOCCCCCCCCCCCCO
Molecular Weight (g/mol)202.34
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Flakes or pellets
Melting point79°C to 83°C
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only