Invitrogen™

SYBR GreenER™ qPCR SuperMix Universal

Catalog number: 11762500
Have Questions?
Invitrogen™

SYBR GreenER™ qPCR SuperMix Universal

Catalog number: 11762500
Have Questions?
Catalog Number
11762500
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2,000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
11762500500 reactions
-
1176202K2,000 reactions
-
11762100100 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of SYBR GreenER qPCR SuperMix and added additional capabilities for your gene expression analysis.

**********************
SYBR™ GreenER™ qPCR SuperMixes Universal incorporate the latest high-performance technology to provide the most reliable gene expression data on a variety of platforms, including ABI 7500, Corbett Rotor-Gene™, MJ Chromo4™ and Opticon™, and Stratagene instruments. The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility with:
• A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings
• An optimized buffer system - improves sensitivity and provides excellent long-term stability
• Uracil DNA glycosylase - reduces carryover contamination in qPCR

Novel Formulation Provides More Reliable Data
The SYBR™ GreenER™ qPCR SuperMix kits include a new dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.

Consistent Specificity and Low-Copy Detection
The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.

Eight cDNA targets and eight genomic DNA targets were amplified from 1 ng of cDNA and 6 pg of gDNA, respectively, on the ABI PRISM™ 7900HT Sequence Detection System using the SYBR™ GreenER™ qPCR SuperMix for ABI PRISM™ instruments. For all 16 targets, detection was positive in the presence of template but negative for all no-template controls.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
SYBR
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Reaction Speed
Standard
For Use With (Equipment)
7500 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P, MJ Chromo4, MJ Opticon
Product Line
Platinum™, SYBR™ GreenER™
Product Type
Real Time PCR SYBR Master Mix
Shipping Condition
Dry Ice
Sufficient For
500 Reactions of 50 μL
For Use With (Application)
Gene Expression
Concentration
2X
No. of Reactions
500 Reactions
Sample Type
DNA (Genomic), cDNA

Contents & Storage

Contains SYBR™ GreenER™ qPCR SuperMix Universal (12.5 mL). SYBR™ GreenER™ qPCR SuperMix Universal is a 2X SuperMix with ROX Reference dye provided in a separate tube (500 μL). Sufficient reagents are provided for 500 reactions based on a 50 μL reaction size.

Store at +4°C upon receipt.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text