Thermo Scientific Chemicals

Eicosanoic acid, 99%

Catalog number: 117720250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Eicosanoic acid, 99%

Catalog number: 117720250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 117720250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
506-30-9
IUPAC Name
icosanoic acid
Molecular Formula
C20H40O2
InChI Key
VKOBVWXKNCXXDE-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCC(O)=O
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Titration with TBAH
97.5 to 102.5 %
Appearance (Form)
Crystalline powder or crystals or flakes
Melting point
73°C to 78°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text