Elaidic acid, 98%, Thermo Scientific Chemicals
Elaidic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Elaidic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
View all
Glass bottle
Catalog number 117730010
Price (USD)/ Each
73.40
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
73.40
Add to cart
Elaidic acid, 98%, Thermo Scientific Chemicals
Catalog number117730010
Price (USD)/ Each
73.40
-
Add to cart
Chemical Identifiers
CAS112-79-8
IUPAC Name(9E)-octadec-9-enoic acid
Molecular FormulaC18H34O2
InChI KeyZQPPMHVWECSIRJ-MDZDMXLPSA-N
SMILESCCCCCCCCC=CCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Powder or crystalline powder or flakes
Titration with NaOH97.5 to 102.5 %
Melting point41°C to 46°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only