Thermo Scientific Chemicals

Epichlorohydrin, 99%, Thermo Scientific Chemicals

Catalog number: 117780100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Epichlorohydrin, 99%, Thermo Scientific Chemicals

Catalog number: 117780100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 117780100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
106-89-8
IUPAC Name
2-(chloromethyl)oxirane
Molecular Formula
C3H5ClO
InChI Key
BRLQWZUYTZBJKN-UHFFFAOYNA-N
SMILES
ClCC1CO1
Color scale
=<15 APHA
Infrared spectrum
Conforms
Water
=<0.02 % (K.F.)
Appearance (Form)
Clear liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text