meso-Erythritol, 99%
meso-Erythritol, 99%
meso-Erythritol, 99%
meso-Erythritol, 99%
Thermo Scientific Chemicals

meso-Erythritol, 99%

CAS: 149-32-6 | C4H10O4 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
25 g
Packaging:
View all
Plastic bottle
Catalog number 117825000
Price (USD)
479.00
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
479.00
Each
Add to cart
Chemical Identifiers
CAS149-32-6
IUPAC Namebutane-1,2,3,4-tetrol
Molecular FormulaC4H10O4
InChI KeyUNXHWFMMPAWVPI-UHFFFAOYNA-N
SMILESOCC(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 % (ELSD)
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals
Water=<1 % (K.F.)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection